Assay Details
| Gene Symbol: FIP1[V] | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345286.1 | NP_001332116.1 | 3 - 4 | 1249 | 68 | |
| NM_125189.3 | NP_200612.2 | 3 - 4 | 1246 | 68 | ||
More Information
Set Membership: |
|

FIP1[V]
Genomic Map