Assay Details
| Gene Symbol: FIP1[V] | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125189.3 | NP_200612.2 | 8 - 9 | 3645 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FIP1[V]| Gene Symbol: FIP1[V] | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125189.3 | NP_200612.2 | 8 - 9 | 3645 | 69 | |
Set Membership: |
|