Assay Details
| Gene Symbol: SNX2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125230.6 | NP_200652.2 | 4 - 5 | 1956 | 80 | |
| GenBank mRNA | AY099775.1 | - | 4 - 5 | 1739 | 80 | |
| BT000285.1 | - | 4 - 5 | 1675 | 80 | ||
More Information
Set Membership: |
|

SNX2a
Genomic Map