Assay Details
| Gene Symbol: ATL63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125245.2 | NP_200666.1 | 1 | 1018 | 108 | |
| GenBank mRNA | DQ056726.1 | - | 1 | 848 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATL63| Gene Symbol: ATL63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125245.2 | NP_200666.1 | 1 | 1018 | 108 | |
| GenBank mRNA | DQ056726.1 | - | 1 | 848 | 108 | |
Set Membership: |
|