Assay Details
| Gene Symbol: PLC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345317.1 | NP_001332555.1 | 3 - 4 | 982 | 128 | |
| NM_001345318.1 | NP_001332556.1 | 3 - 4 | 675 | 128 | ||
| NM_125256.3 | NP_200677.2 | 2 - 3 | 1056 | 128 | ||
| GenBank mRNA | AF434167.2 | - | 2 - 3 | 619 | 128 | |
| AY062681.1 | - | 2 - 3 | 629 | 128 | ||
| BT010399.1 | - | 2 - 3 | 528 | 128 | ||
More Information
Set Membership: |
|

PLC5
Genomic Map