Assay Details
| Gene Symbol: PLC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345317.1 | NP_001332555.1 | 5 - 6 | 1310 | 126 | |
| NM_001345318.1 | NP_001332556.1 | 5 - 6 | 1003 | 126 | ||
| NM_125256.3 | NP_200677.2 | 4 - 5 | 1384 | 126 | ||
| GenBank mRNA | AF434167.2 | - | 4 - 5 | 947 | 126 | |
| AY062681.1 | - | 4 - 5 | 957 | 126 | ||
| BT010399.1 | - | 4 - 5 | 856 | 126 | ||
More Information
Set Membership: |
|

PLC5
Genomic Map