Assay Details
| Gene Symbol: PLC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345317.1 | NP_001332555.1 | 6 - 7 | 1535 | 98 | |
| NM_001345318.1 | NP_001332556.1 | 6 - 7 | 1228 | 98 | ||
| NM_125256.3 | NP_200677.2 | 5 - 6 | 1609 | 98 | ||
| GenBank mRNA | AF434167.2 | - | 5 - 6 | 1172 | 98 | |
| AY062681.1 | - | 5 - 6 | 1182 | 98 | ||
| BT010399.1 | - | 5 - 6 | 1081 | 98 | ||
More Information
Set Membership: |
|

PLC5
Genomic Map