Assay Details
| Gene Symbol: PLC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345317.1 | NP_001332555.1 | 8 - 9 | 1897 | 107 | |
| NM_001345318.1 | NP_001332556.1 | 8 - 9 | 1590 | 107 | ||
| NM_125256.3 | NP_200677.2 | 7 - 8 | 1971 | 107 | ||
| GenBank mRNA | AF434167.2 | - | 7 - 8 | 1534 | 107 | |
| AY062681.1 | - | 7 - 8 | 1544 | 107 | ||
| BT010399.1 | - | 7 - 8 | 1443 | 107 | ||
More Information
Set Membership: |
|

PLC5
Genomic Map