Assay Details
| Gene Symbol: PLC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037020.2 | NP_001032097.1 | 2 - 3 | 320 | 142 | |
| NM_001345319.1 | NP_001318832.1 | 3 - 4 | 714 | 142 | ||
| NM_125257.3 | NP_200678.2 | 2 - 3 | 777 | 142 | ||
| GenBank mRNA | AF434168.2 | - | 2 - 3 | 525 | 142 | |
| AK317146.1 | - | 2 - 3 | 321 | 142 | ||
| AY093217.1 | - | 2 - 3 | 566 | 142 | ||
| BT008358.1 | - | 2 - 3 | 525 | 142 | ||
More Information
Set Membership: |
|

PLC4
Genomic Map