Assay Details
| Gene Symbol: PLC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037020.2 | NP_001032097.1 | 6 - 7 | 1056 | 64 | |
| NM_001345319.1 | NP_001318832.1 | 7 - 8 | 1450 | 64 | ||
| NM_125257.3 | NP_200678.2 | 6 - 7 | 1513 | 64 | ||
| GenBank mRNA | AF434168.2 | - | 6 - 7 | 1261 | 64 | |
| AK317146.1 | - | 6 - 7 | 1057 | 64 | ||
| AY093217.1 | - | 6 - 7 | 1302 | 64 | ||
| BT008358.1 | - | 6 - 7 | 1261 | 64 | ||
More Information
Set Membership: |
|

PLC4
Genomic Map