Assay Details
| Gene Symbol: UXS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085299.1 | NP_001078768.1 | 10 - 11 | 887 | 101 | |
| NM_001345342.1 | NP_001331974.1 | 10 - 11 | 956 | 101 | ||
| NM_001345343.1 | NP_001331973.1 | 11 - 12 | 1256 | 101 | ||
| NM_125319.3 | NP_200737.1 | 11 - 12 | 1062 | 101 | ||
| GenBank mRNA | AF387789.1 | - | 10 - 11 | 842 | 101 | |
| AY088443.1 | - | 11 - 12 | 988 | 101 | ||
More Information
Set Membership: |
|

UXS3
Genomic Map