Assay Details
| Gene Symbol: UXS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085299.1 | NP_001078768.1 | 5 - 6 | 455 | 73 | |
| NM_001345342.1 | NP_001331974.1 | 5 - 6 | 524 | 73 | ||
| NM_001345343.1 | NP_001331973.1 | 6 - 7 | 824 | 73 | ||
| NM_125319.3 | NP_200737.1 | 6 - 7 | 630 | 73 | ||
| GenBank mRNA | AF387789.1 | - | 5 - 6 | 410 | 73 | |
More Information
Set Membership: |
|

UXS3
Genomic Map