Assay Details
| Gene Symbol: AAK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125427.3 | NP_200842.2 | 1 - 2 | 440 | 68 | |
| GenBank mRNA | AK117598.1 | - | 1 - 2 | 262 | 68 | |
| AY079019.1 | - | 1 - 2 | 325 | 68 | ||
| AY093748.1 | - | 1 - 2 | 209 | 68 | ||
More Information
Set Membership: |
|

AAK6
Genomic Map