Assay Details
| Gene Symbol: ARF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345418.1 | NP_001331788.1 | 8 - 9 | 1601 | 93 | |
| NM_125438.4 | NP_200853.1 | 7 - 8 | 1673 | 93 | ||
| GenBank mRNA | AF013466.1 | - | 7 - 8 | 1131 | 93 | |
| AF344313.1 | - | 7 - 8 | 1076 | 93 | ||
| AY080832.1 | - | 7 - 8 | 1433 | 93 | ||
| AY113977.1 | - | 7 - 8 | 1076 | 93 | ||
More Information
Set Membership: |
|

ARF4
Genomic Map