Assay Details
| Gene Symbol: GPAT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125455.4 | NP_568925.1 | 11 - 12 | 1203 | 58 | |
| GenBank mRNA | AK118869.1 | - | 11 - 12 | 1129 | 58 | |
| BT006033.1 | - | 11 - 12 | 1079 | 58 | ||
| FJ479752.1 | - | 11 - 12 | 1079 | 58 | ||
More Information
Set Membership: |
|

GPAT9
Genomic Map