Assay Details
| Gene Symbol: COB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125485.4 | NP_568930.1 | 1 - 2 | 514 | 114 | |
| GenBank mRNA | AY094402.1 | - | 1 - 2 | 215 | 114 | |
| BT001119.1 | - | 1 - 2 | 108 | 114 | ||
More Information
Set Membership: |
|

COB
Genomic Map