Assay Details
| Gene Symbol: COB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345435.1 | NP_001330939.1 | 3 - 4 | 726 | 134 | |
| NM_125485.4 | NP_568930.1 | 3 - 4 | 710 | 134 | ||
| GenBank mRNA | AY094402.1 | - | 3 - 4 | 411 | 134 | |
| BT001119.1 | - | 3 - 4 | 304 | 134 | ||
More Information
Set Membership: |
|

COB
Genomic Map