Assay Details
| Gene Symbol: AT5G60990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125492.5 | NP_568931.1 | 7 - 8 | 858 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G60990| Gene Symbol: AT5G60990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125492.5 | NP_568931.1 | 7 - 8 | 858 | 87 | |
Set Membership: |
|