Assay Details
| Gene Symbol: AT5G61050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125498.1 | NP_200913.1 | 4 - 5 | 597 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G61050| Gene Symbol: AT5G61050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125498.1 | NP_200913.1 | 4 - 5 | 597 | 129 | |
Set Membership: |
|