Assay Details
| Gene Symbol: AT5G61450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125538.3 | NP_200953.1 | 11 - 12 | 1221 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G61450| Gene Symbol: AT5G61450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125538.3 | NP_200953.1 | 11 - 12 | 1221 | 96 | |
Set Membership: |
|