Assay Details
| Gene Symbol: MIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125539.2 | NP_200954.1 | 21 - 22 | 2738 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIM| Gene Symbol: MIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125539.2 | NP_200954.1 | 21 - 22 | 2738 | 113 | |
Set Membership: |
|