Assay Details
| Gene Symbol: MIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125539.2 | NP_200954.1 | 9 - 10 | 1319 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIM| Gene Symbol: MIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125539.2 | NP_200954.1 | 9 - 10 | 1319 | 83 | |
Set Membership: |
|