Assay Details
| Gene Symbol: AT5G61970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125590.3 | NP_568947.1 | 10 - 11 | 1418 | 119 | |
| GenBank mRNA | AY051002.1 | - | 10 - 11 | 1348 | 119 | |
| AY079306.1 | - | 9 - 10 | 1199 | 119 | ||
More Information
Set Membership: |
|

AT5G61970
Genomic Map