Assay Details
| Gene Symbol: AT5G61970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125590.3 | NP_568947.1 | 3 - 4 | 380 | 79 | |
| GenBank mRNA | AY051002.1 | - | 3 - 4 | 310 | 79 | |
| AY079306.1 | - | 2 - 3 | 161 | 79 | ||
More Information
Set Membership: |
|

AT5G61970
Genomic Map