Assay Details
| Gene Symbol: AT5G61970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125590.3 | NP_568947.1 | 5 - 6 | 687 | 105 | |
| GenBank mRNA | AY051002.1 | - | 5 - 6 | 617 | 105 | |
| AY079306.1 | - | 4 - 5 | 468 | 105 | ||
More Information
Set Membership: |
|

AT5G61970
Genomic Map