Assay Details
| Gene Symbol: AT5G61970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125590.3 | NP_568947.1 | 6 - 7 | 778 | 101 | |
| GenBank mRNA | AY051002.1 | - | 6 - 7 | 708 | 101 | |
| AY079306.1 | - | 5 - 6 | 559 | 101 | ||
More Information
Set Membership: |
|

AT5G61970
Genomic Map