Assay Details
| Gene Symbol: AT5G62080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125601.3 | NP_568949.1 | 1 - 2 | 357 | 92 | |
| GenBank mRNA | AK117876.1 | - | 1 - 2 | 316 | 92 | |
More Information
Set Membership: |
|

AT5G62080
Genomic Map