Assay Details
| Gene Symbol: AT5G62140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125607.4 | NP_201020.1 | 1 - 2 | 570 | 90 | |
| GenBank mRNA | BT029994.1 | - | 1 - 2 | 443 | 90 | |
More Information
Set Membership: |
|

AT5G62140
Genomic Map