Assay Details
| Gene Symbol: ZIP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125609.2 | NP_201022.1 | 1 - 2 | 615 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZIP12| Gene Symbol: ZIP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125609.2 | NP_201022.1 | 1 - 2 | 615 | 149 | |
Set Membership: |
|