Assay Details
| Gene Symbol: PRH75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125613.3 | NP_201025.1 | 2 - 3 | 733 | 71 | |
| GenBank mRNA | AY056137.1 | - | 2 - 3 | 659 | 71 | |
| BT008581.1 | - | 2 - 3 | 568 | 71 | ||
More Information
Set Membership: |
|

PRH75
Genomic Map