Assay Details
| Gene Symbol: CDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125637.4 | NP_201049.3 | 2 | 1108 | 115 | |
| GenBank mRNA | AB231333.1 | - | 2 | 784 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDF1| Gene Symbol: CDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125637.4 | NP_201049.3 | 2 | 1108 | 115 | |
| GenBank mRNA | AB231333.1 | - | 2 | 784 | 115 | |
Set Membership: |
|