Assay Details
| Gene Symbol: EB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125644.3 | NP_201056.1 | 5 - 6 | 791 | 63 | |
| GenBank mRNA | AY084598.1 | - | 5 - 6 | 684 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EB1B| Gene Symbol: EB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125644.3 | NP_201056.1 | 5 - 6 | 791 | 63 | |
| GenBank mRNA | AY084598.1 | - | 5 - 6 | 684 | 63 | |
Set Membership: |
|