Assay Details
| Gene Symbol: AT5G62550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125649.5 | NP_201061.1 | 1 - 2 | 184 | 75 | |
| GenBank mRNA | AK229459.1 | - | 1 - 2 | 122 | 75 | |
More Information
Set Membership: |
|

AT5G62550
Genomic Map