Assay Details
| Gene Symbol: AT5G62550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125649.5 | NP_201061.1 | 2 - 3 | 293 | 109 | |
| GenBank mRNA | AK229459.1 | - | 2 - 3 | 231 | 109 | |
More Information
Set Membership: |
|

AT5G62550
Genomic Map