Assay Details
| Gene Symbol: AT5G62550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125649.5 | NP_201061.1 | 3 - 4 | 667 | 112 | |
| GenBank mRNA | AK229459.1 | - | 3 - 4 | 605 | 112 | |
More Information
Set Membership: |
|

AT5G62550
Genomic Map