Assay Details
| Gene Symbol: AT1G49230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103813.4 | NP_175349.1 | 1 | 731 | 79 | |
| GenBank mRNA | AK176235.1 | - | 1 | 664 | 79 | |
| AK176512.1 | - | 1 | 635 | 79 | ||
| BT010489.1 | - | 1 | 605 | 79 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

AT1G49230
Genomic Map