Assay Details
| Gene Symbol: AT1G49250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103816.2 | NP_175351.1 | 2 - 3 | 505 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G49250| Gene Symbol: AT1G49250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103816.2 | NP_175351.1 | 2 - 3 | 505 | 110 | |
Set Membership: |
|