Assay Details
| Gene Symbol: ICK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333395.1 | NP_001322045.1 | 3 - 4 | 705 | 73 | |
| NM_103850.2 | NP_175385.1 | 3 - 4 | 520 | 73 | ||
More Information
Set Membership: |
|

ICK5
Genomic Map