Assay Details
| Gene Symbol: PREP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103851.3 | NP_175386.2 | 19 - 20 | 3353 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PREP2| Gene Symbol: PREP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103851.3 | NP_175386.2 | 19 - 20 | 3353 | 106 | |
Set Membership: |
|