Assay Details
| Gene Symbol: ABF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333400.1 | NP_001322564.1 | 3 - 4 | 1209 | 113 | |
| NM_103859.5 | NP_564551.1 | 3 - 4 | 1314 | 113 | ||
| GenBank mRNA | AF093544.1 | - | 2 - 3 | 1147 | 113 | |
| AK226983.1 | - | 3 - 4 | 1226 | 113 | ||
| BT008575.1 | - | 2 - 3 | 1072 | 113 | ||
| BT008647.1 | - | 3 - 4 | 1225 | 113 | ||
More Information
Set Membership: |
|

ABF1
Genomic Map