Assay Details
| Gene Symbol: PAB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198255.1 | NP_001185184.1 | 4 - 5 | 1416 | 87 | |
| NM_103863.3 | NP_564554.1 | 4 - 5 | 1415 | 87 | ||
| GenBank mRNA | AF370517.1 | - | 4 - 5 | 1410 | 87 | |
| AY064622.1 | - | 4 - 5 | 1150 | 87 | ||
More Information
Set Membership: |
|

PAB8
Genomic Map