Assay Details
| Gene Symbol: CLPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103884.4 | NP_564560.1 | 2 - 3 | 543 | 137 | |
| GenBank mRNA | AB022330.1 | - | 2 - 3 | 498 | 137 | |
| AY045677.1 | - | 2 - 3 | 455 | 137 | ||
| BT000572.1 | - | 2 - 3 | 351 | 137 | ||
More Information
Set Membership: |
|

CLPR1
Genomic Map