Assay Details
| Gene Symbol: CLPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103884.4 | NP_564560.1 | 6 - 7 | 1029 | 122 | |
| GenBank mRNA | AB022330.1 | - | 6 - 7 | 984 | 122 | |
| AY045677.1 | - | 6 - 7 | 941 | 122 | ||
| BT000572.1 | - | 6 - 7 | 837 | 122 | ||
More Information
Set Membership: |
|

CLPR1
Genomic Map