Assay Details
| Gene Symbol: TOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103891.4 | NP_175425.2 | 36 - 37 | 5033 | 82 | |
| GenBank mRNA | AF178967.1 | - | 36 - 37 | 4888 | 82 | |
More Information
Set Membership: |
|

TOR
Genomic Map