Assay Details
| Gene Symbol: TOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123987.1 | NP_001117459.1 | 42 - 43 | 5919 | 90 | |
| NM_103891.4 | NP_175425.2 | 42 - 43 | 6000 | 90 | ||
| GenBank mRNA | AF178967.1 | - | 42 - 43 | 5855 | 90 | |
More Information
Set Membership: |
|

TOR
Genomic Map