Assay Details
| Gene Symbol: TOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123987.1 | NP_001117459.1 | 55 - 56 | 7723 | 69 | |
| NM_103891.4 | NP_175425.2 | 55 - 56 | 7804 | 69 | ||
More Information
Set Membership: |
|

TOR
Genomic Map