Assay Details
| Gene Symbol: DWF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084224.1 | NP_001077693.1 | 2 - 3 | 403 | 75 | |
| NM_103926.5 | NP_175460.1 | 2 - 3 | 405 | 75 | ||
| GenBank mRNA | AK227279.1 | - | 2 - 3 | 407 | 75 | |
| AY099589.1 | - | 2 - 3 | 359 | 75 | ||
| BT000245.1 | - | 2 - 3 | 308 | 75 | ||
More Information
Set Membership: |
|

DWF5
Genomic Map