Assay Details
| Gene Symbol: DWF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103926.5 | NP_175460.1 | 3 - 4 | 462 | 77 | |
| GenBank mRNA | AK227279.1 | - | 3 - 4 | 464 | 77 | |
| AY099589.1 | - | 3 - 4 | 416 | 77 | ||
| BT000245.1 | - | 3 - 4 | 365 | 77 | ||
More Information
Set Membership: |
|

DWF5
Genomic Map