Assay Details
| Gene Symbol: DWF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084224.1 | NP_001077693.1 | 5 - 6 | 627 | 93 | |
| NM_103926.5 | NP_175460.1 | 5 - 6 | 632 | 93 | ||
| GenBank mRNA | AK227279.1 | - | 5 - 6 | 634 | 93 | |
| AY099589.1 | - | 5 - 6 | 586 | 93 | ||
| BT000245.1 | - | 5 - 6 | 535 | 93 | ||
More Information
Set Membership: |
|

DWF5
Genomic Map