Assay Details
| Gene Symbol: HIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198259.1 | NP_001185188.1 | 6 - 7 | 621 | 96 | |
| NM_001333440.1 | NP_001322424.1 | 5 - 6 | 851 | 96 | ||
| NM_001333441.1 | NP_001322422.1 | 6 - 7 | 650 | 96 | ||
| NM_001333442.1 | NP_001322423.1 | 6 - 7 | 606 | 96 | ||
| NM_103933.6 | NP_564573.3 | 6 - 7 | 747 | 96 | ||
| GenBank mRNA | AK228745.1 | - | 6 - 7 | 602 | 96 | |
More Information
Set Membership: |
|

HIT1
Genomic Map